missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDRG1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-00250-50ul
This item is not returnable.
View return policy
Description
PDRG1 Polyclonal antibody specifically detects PDRG1 in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin)
Specifications
| PDRG1 | |
| Polyclonal | |
| ELISA 1:5000-1:10000, Immunohistochemistry-Paraffin 1:2500-1:10000 | |
| C20orf126p53 and DNA damage regulated 1, chromosome 20 open reading frame 126, dJ310O13.3, p53 and DNA-damage regulated 1, PDRGp53 and DNA damage-regulated protein 1 | |
| Produced in rabbits immunized with purified, recombinant Human PDRG1 (Uniprot#: Q9NUG6; Met1-Gly133) | |
| 50 μg | |
| Primary | |
| Human | |
| Purified |
| ELISA, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 81572 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction