missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PDP2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5100680
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total PDP2.
Catalyzes the dephosphorylation and concomitant reactivation of the alpha subunit of the E1 component of the pyruvate dehydrogenase complex.
Specifications
| PDP2 | |
| Polyclonal | |
| Unconjugated | |
| Pdp2 | |
| [pyruvate dehydrogenase (acetyl-transferring)]-phosphatase; [Pyruvate dehydrogenase [acetyl-transferring]]-phosphatase 2, mitochondrial; 4833426J09Rik; Gm1705; KIAA1348; mKIAA1348; PDP 2; PDP2; PDPC 2; PPM2B; PPM2C2; protein phosphatase 2C, magnesium-dependent, catalytic subunit 2; protein phosphatase, Mg2+/Mn2+ dependent 2B; pyruvate dehydrogenase [acetyl-transferring]-phosphatase 2, mitochondrial; pyruvate dehydrogenase phosphatase catalytic subunit 2; pyruvate dehydrogenase phosphatase isoenzyme 2; pyruvate dehydrogenase phosphatase, catalytic subunit 2; pyruvate dehyrogenase phosphatase catalytic subunit 2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 246311, 382051, 57546 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O88484, Q9P2J9 | |
| Pdp2 | |
| A synthesized peptide derived from human PDP2(Accession Q9P2J9), corresponding to amino acid residues M1-R44. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction