missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDP2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-82912-25ul
This item is not returnable.
View return policy
Description
PDP2 Polyclonal specifically detects PDP2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| PDP2 | |
| Polyclonal | |
| Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| EC 3.1.3, EC 3.1.3.43, KIAA1348[Pyruvate dehydrogenase [acetyl-transferring]]-phosphatase 2, mitochondrial, PDP 2, PDPC 2, PPM2C2, protein phosphatase 2C, magnesium-dependent, catalytic subunit 2, Pyruvate dehydrogenase phosphatase catalytic subunit 2, pyruvate dehydrogenase phosphatase isoenzyme 2, pyruvate dehydrogenase phosphatase, catalytic subunit 2, pyruvate dehyrogenase phosphatase catalytic subunit 2 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| PDP2 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:DMLSNEDVVRLVVGHLAEADWHKTDLAQRPANLGLMQSLLLQRKASGLHEADQNAATRLIRHAIGNNEYGE | |
| 25 μL | |
| Protein Phosphatase | |
| 57546 | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction