missing translation for 'onlineSavingsMsg'
Learn More

PDP1/PPAPDC2 Antibody, DyLight 755, Novus Biologicals™

Product Code. 18428782 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18428782 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18428782 Supplier Novus Biologicals Supplier No. NBP176245IR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PDP1/PPAPDC2 Polyclonal antibody specifically detects PDP1/PPAPDC2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen PDP1/PPAPDC2
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate DyLight 755
Formulation 50mM Sodium Borate
Gene Alias bA6J24.6, EC 3.1.3, EC 3.1.3.-, FLJ46512, FLJ90191, MGC15483, PDP1, phosphatidic acid phosphatase type 2 domain containing 2, Phosphatidic acid phosphatase type 2 domain-containing protein 2, polyisoprenoid diphosphate phosphatase type 1, PPAP2 domain-containing protein 2, presqualene diphosphate phosphatase, PSDP
Host Species Rabbit
Immunogen Antibody was raised against a 13 amino acid synthetic peptide from near the carboxy terminus human PPAPDC2. The immunogen is located within amino acids 170 - 220 of PPAPDC2. Amino Acid Squence: LVRRRRPAHNNQMD
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 403313
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.