missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PDP1 Polyclonal Antibody

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA553815

Product Code. 15741751

  • £467.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: YLTPPQVNSI LKANEYSFKV PEFDGKNVSS ILGFDSNQLP ANAPIEDRRS AATCLQTRGM LLGVFDGHAG CACSQ Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

PDP1 is a Pyruvate dehydrogenase and helps catalyze phosphorylation of serine residues of E1 to inactivate the E1 component and inhibit the complex. Pyruvate dehydrogenase phosphatases catalyze the dephosphorylation and activation of the E1 component to reverse the effects of pyruvate dehydrogenase kinases. Pyruvate dehydrogenase phosphatase is a heterodimer consisting of catalytic and regulatory subunits.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PDP1
Polyclonal
Unconjugated
PDP1
[Pyruvate dehydrogenase [acetyl-transferring]]-phosphatase 1, mitochondrial; FLJ32517; FLJ56179; Gm1024; MGC119646; PDH; PDP; PDP 1; PDP1; PDPC; PDPC 1; PPM2A; Ppm2c; Protein phosphatase 2C; protein phosphatase 2C, magnesium dependent, catalytic subunit; protein phosphatase 2C, magnesium-dependent, catalytic subunit; protein phosphatase, Mg2+/Mn2+ dependent 2A; pyruvate dehydrogenase; pyruvate dehydrogenase (Lipoamide) phosphatase-phosphatase; pyruvate dehydrogenase phosphatase catalytic subunit 1; pyruvate dehydrogenase phosphatase isoenzyme 1; pyruvate dehydrogenase phosphatase isoenzyme 2; pyruvate dehydrogenase phosphatase, catalytic subunit 1; pyruvate dehyrogenase phosphatase catalytic subunit 1
Rabbit
Antigen affinity chromatography
RUO
54704
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.10 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q9P0J1
PDP1
Recombinant protein corresponding to Human PDP1. Recombinant protein control fragment (Product #RP-91576).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.