missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDLIM5 Antibody (OTI1H10), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73317PCP
This item is not returnable.
View return policy
Description
PDLIM5 Monoclonal specifically detects PDLIM5 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.
Specifications
| PDLIM5 | |
| Monoclonal | |
| PerCP | |
| PBS with 0.05% Sodium Azide | |
| PDLIM5 | |
| Full length recombinant protein of human PDLIM5 (NP_006448) produced in HEK293T cells. | |
| 0.1 mL | |
| 10611.0 | |
| Store at 4°C in the dark. |
| Flow Cytometry | |
| OTI1H10 | |
| Flow Cytometry | |
| Enh, ENH1, ENHLIM, Enigma homolog, enigma-like LIM domain protein, Enigma-like PDZ and LIM domains protein, PDZ and LIM domain 5, PDZ and LIM domain protein 5 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction