missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDLIM5 Antibody (OTI1H10), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-01877
This item is not returnable.
View return policy
Description
PDLIM5 Monoclonal specifically detects PDLIM5 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry.
Specifications
| PDLIM5 | |
| Monoclonal | |
| 3.24 mg/mL | |
| Western Blot 1:2000, Flow Cytometry 1:100 | |
| PDLIM5 | |
| Full length human recombinant protein of human PDLIM5(NP_006448) produced in HEK293T cell. | |
| Affinity Purified | |
| RUO | |
| 10611 | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot, Flow Cytometry | |
| OTI1H10 | |
| Unconjugated | |
| Enh, ENH1, ENHLIM, Enigma homolog, enigma-like LIM domain protein, Enigma-like PDZ and LIM domains protein, PDZ and LIM domain 5, PDZ and LIM domain protein 5 | |
| Mouse | |
| 63.8 kDa | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction