missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDLIM5 Antibody (OTI1B5), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-73316CL1
This item is not returnable.
View return policy
Description
PDLIM5 Monoclonal antibody specifically detects PDLIM5 in Human,Mouse,Rat samples. It is validated for Western Blot
Specifications
| PDLIM5 | |
| Monoclonal | |
| CoraFluor 1 | |
| Enh, ENH1, ENHLIM, Enigma homolog, enigma-like LIM domain protein, Enigma-like PDZ and LIM domains protein, PDZ and LIM domain 5, PDZ and LIM domain protein 5 | |
| Full length human recombinant protein of human PDLIM5(NP_006448) produced in HEK293T cell. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| OTI1B5 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| 10611 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction