missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDLIM3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13199-2-AP-150UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene contains a PDZ domain and a LIM domain, indicating that it may be involved in cytoskeletal assembly. In support of this, the encoded protein has been shown to bind the spectrin-like repeats of alpha-actinin-2 and to colocalize with alpha-actinin-2 at the Z lines of skeletal muscle. This gene is found near a region of chromosome 4 that has been implicated in facioscapulohumeral muscular dystrophy, but this gene does not appear to be involved in the disease. Two transcript variants encoding different isoforms have been found for this gene.Specifications
| PDLIM3 | |
| Polyclonal | |
| Unconjugated | |
| PDLIM3 | |
| Actinin associated LIM protein, ALP, PDLIM3, PDZ and LIM domain 3, PDZ and LIM domain protein 3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 114108, 27295, 53318 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.41 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O70209, Q53GG5, Q66HS7 | |
| PDLIM3 | |
| PDLIM3 Fusion Protein Ag3934 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction