Learn More
PDLIM3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 13199-2-AP-20UL
 View more versions of this product
Product Code. 16801304
  • £135.00 / 20µL
Description
The protein encoded by this gene contains a PDZ domain and a LIM domain, indicating that it may be involved in cytoskeletal assembly. In support of this, the encoded protein has been shown to bind the spectrin-like repeats of alpha-actinin-2 and to colocalize with alpha-actinin-2 at the Z lines of skeletal muscle. This gene is found near a region of chromosome 4 that has been implicated in facioscapulohumeral muscular dystrophy, but this gene does not appear to be involved in the disease. Two transcript variants encoding different isoforms have been found for this gene.
Specifications
Specifications
PDLIM3
Polyclonal
Unconjugated
PDLIM3
Actinin associated LIM protein, ALP, PDLIM3, PDZ and LIM domain 3, PDZ and LIM domain protein 3
Rabbit
Antigen Affinity Chromatography
RUO
114108, 27295, 53318
-20°C
Liquid
Western Blot
0.41 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O70209, Q53GG5, Q66HS7
PDLIM3
PDLIM3 Fusion Protein Ag3934
20 μL
Primary
Rat, Mouse, Human