missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDLIM2 Antibody (OTI11A12), DyLight 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73311V
This item is not returnable.
View return policy
Description
PDLIM2 Monoclonal specifically detects PDLIM2 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| PDLIM2 | |
| Monoclonal | |
| DyLight 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| PDLIM2 | |
| Full length recombinant protein of human PDLIM2 (NP_067643) produced in HEK293T cells. | |
| 0.1 mL | |
| 64236.0 | |
| Store at 4°C in the dark. |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI11A12 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| FLJ34715, MYSTIQUE, PDZ and LIM domain 2 (mystique), PDZ and LIM domain protein 2, PDZ-LIM protein mystique, SLIM | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction