missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PDLIM1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5112292
This item is not returnable.
View return policy
Description
PDLIM1 Polyclonal Antibody for Western Blot, IP
Initiation of transcription by RNA polymerase II requires the activities of more than 70 polypeptides. The protein that coordinates these activities is transcription factor IID (TFIID), which binds to the core promoter to position the polymerase properly, serves as the scaffold for assembly of the remainder of the transcription complex, and acts as a channel for regulatory signals. TFIID is composed of the TATA-binding protein (TBP) and a group of evolutionarily conserved proteins known as TBP-associated factors or TAFs. TAFs may participate in basal transcription, serve as coactivators, function in promoter recognition or modify general transcription factors (GTFs) to facilitate complex assembly and transcription initiation. A distinctive feature of TBP is a long string of glutamines in the N-terminal. This region of the protein modulates the DNA binding activity of the C terminus, and modulation of DNA binding affects the rate of transcription complex formation and initiation of transcription. Mutations that expand the number of CAG repeats encoding this polyglutamine tract, and thus increase the length of the polyglutamine string, are associated with spinocerebellar ataxia 17, a neurodegenerative disorder classified as a polyglutamine disease.
Specifications
| PDLIM1 | |
| Polyclonal | |
| Unconjugated | |
| Pdlim1 | |
| carboxyl terminal LIM domain protein 1; Clim1; Clp36; CLP-36; C-terminal LIM domain protein 1; elfin; enigma; epididymis secretory protein Li 112; hCLIM1; HEL-S-112; LIM domain protein CLP-36; LIM protein; mClim1; Pdlim1; PDZ and LIM domain 1; PDZ and LIM domain 1 (elfin); PDZ and LIM domain protein 1 | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| 9124 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 1 mg/mL | |
| PBS with 0.03% ProClin 300; pH 7 | |
| O00151 | |
| Pdlim1 | |
| E. coli-derived Human PDLIM1 fragment. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction