missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDK3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12215-1-AP-150UL
This item is not returnable.
View return policy
Description
PDK3 inhibits the mitochondrial pyruvate dehydrogenase complex by phosphorylation of the E1 alpha subunit, thus contributing to the regulation of glucose metabolism.Specifications
| PDK3 | |
| Polyclonal | |
| Unconjugated | |
| PDK3 | |
| PDK3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 236900, 5165 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.35 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q15120, Q922H2 | |
| PDK3 | |
| PDK3 Fusion Protein Ag2879 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction