missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PDK3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA528019
This item is not returnable.
View return policy
Description
Recommended positive controls: HepG2, mouse liver, PDK3-transfected 293T. Predicted reactivity: Mouse (96%), Rat (96%), Chicken (93%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
PDK3 inhibits the mitochondrial pyruvate dehydrogenase complex by phosphorylation of the E1 alpha subunit, thus contributing to the regulation of glucose metabolism.
Specifications
| PDK3 | |
| Polyclonal | |
| Unconjugated | |
| PDK3 | |
| [Pyruvate dehydrogenase (acetyl-transferring)] kinase isozyme 3, mitochondrial; [Pyruvate dehydrogenase [lipoamide]] kinase isozyme 3, mitochondrial; 2610001M10Rik; AI035637; CMTX6; GS1-358P8.4; PDHK3; PDK3; pyruvate dehydrogenase; Pyruvate dehydrogenase kinase 3; Pyruvate dehydrogenase kinase isoform 3; pyruvate dehydrogenase kinase, isoenzyme 3; pyruvate dehydrogenase kinase, isozyme 3; pyruvate dehydrogenase, lipoamide, kinase isozyme 3, mitochondrial | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 236900, 5165 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.16 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7 | |
| Q15120, Q922H2 | |
| PDK3 | |
| Recombinant protein encompassing a sequence within the center region of human PDK3. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Amphibian, Human, Mouse, Rodent | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction