missing translation for 'onlineSavingsMsg'
Learn More

PDK3 Antibody, Novus Biologicals™

Product Code. 30231191 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30231191 100 μL 100µL
30231635 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30231191 Supplier Novus Biologicals Supplier No. NBP336720100ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PDK3 Polyclonal antibody specifically detects PDK3 in Human samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen PDK3
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:2000, ELISA
Formulation PBS (pH 7.3), 50% glycerol
Gene Alias EC 2.7.11.2, Pyruvate dehydrogenase kinase isoform 3, pyruvate dehydrogenase kinase, isoenzyme 3, pyruvate dehydrogenase kinase, isozyme 3, pyruvate dehydrogenase, lipoamide, kinase isozyme 3, mitochondrial
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 137-406 of human PDK3 (NP_005382.1).,, Sequence:, FGFDPFISTNIQYFLDRFYTNRISFRMLINQHTLLFGGDTNPVHPKHIGSIDPTCNVADVVKDAYETAKMLCEQYYLVAPELEVEEFNAKAPDKPIQVVYVPSHLFHMLFELFKNSMRATVELYEDRKEGYPAVKTLVTLGKEDLSIKISDLGGGVPLRKIDRLFNYMYSTAPRPSLEPTRAAPLAGFGYGLPISRLYARYFQGDLKLYSMEGVGTDAVIYLKALSSESFERLPVFNKSAWRHYKTTPEADDWSNPSSEPRDASKYKAKQ
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 5165
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.