missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDK2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15647-1-AP-20UL
This item is not returnable.
View return policy
Description
PDK2 inhibits the mitochondrial pyruvate dehydrogenase complex by phosphorylation of the E1 alpha subunit, thus contributing to the regulation of glucose metabolism.Specifications
| PDK2 | |
| Polyclonal | |
| Unconjugated | |
| PDK2 | |
| PDHK2, PDK2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 18604, 5164, 81530 | |
| -20°C | |
| Liquid |
| Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q15119, Q64536, Q9JK42 | |
| PDK2 | |
| PDK2 Fusion Protein Ag8183 | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction