missing translation for 'onlineSavingsMsg'
Learn More

PDK2 Mouse anti-Human, Clone:3F2D7, Abnova™

Mouse monoclonal antibody raised against recombinant human PDK2.

Brand:  Abnova MAB16631.100ug

Product Code. 16074406

  • £535.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Specifications

Specifications

PDK2
Monoclonal
Unconjugated
ELISA (1:10000)
Western Blot (1:500-1:2000)
Flow Cytometry (1:200-1:400)
The optimal working dilution should be determined by the end user.
PDK2
Mouse
100 μg
Human
Antibody
IgG2b
ELISA, Flow Cytometry, Western Blot
3F2D7
pyruvate dehydrogenase kinase, isozyme 2
In PBS (0.05% sodium azide)
PDHK2
Recombinant protein corresponding to amino acid 178-404 of human PDK2 from E. coli.
5164
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.