missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDIK1L Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-19751
This item is not returnable.
View return policy
Description
PDIK1L Polyclonal specifically detects PDIK1L in Human samples. It is validated for Western Blot.
Specifications
| PDIK1L | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal | |
| casein kinase, CLIK1LRP11-96L14.4, EC 2.7.11.1, PDLIM1 interacting kinase 1 like, PDLIM1-interacting kinase 1-like, serine/threonine-protein kinase PDIK1L, STK35L2 | |
| Rabbit | |
| 39 kDa | |
| 0.1 mL | |
| Primary | |
| Human. Species reactivity based on sequence identity: Mouse (100%), Rat (100%), Bovine (100%), Xenopus tropicalis (100%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot | |
| 1mg/mL | |
| Western Blot 1:500-1:3000 | |
| Q8N165 | |
| PDIK1L | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of human PDIK1L. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 149420 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction