missing translation for 'onlineSavingsMsg'
Learn More

PDIA6 Rabbit anti-Human, Mouse, Rat, Clone: 2Q1L1, Novus Biologicals™

Product Code. 18311372 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18311372 20 μg 20µL
18344894 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18311372 Supplier Novus Biologicals Supplier No. NBP31636520UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

PDIA6 Monoclonal antibody specifically detects PDIA6 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen PDIA6
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 2Q1L1
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:200-1:500
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias EC 5.3.4.1, Endoplasmic reticulum protein 5, ERp5, ERP5ER protein 5, P5TXNDC7, protein disulfide isomerase family A, member 6, Protein disulfide isomerase P5, protein disulfide isomerase-associated 6, protein disulfide isomerase-related protein, protein disulfide-isomerase A6, thioredoxin domain containing 7 (protein disulfide isomerase), Thioredoxin domain-containing protein 7
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human PDIA6 (Q15084).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 10130
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.