missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ PDIA6 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-29117
This item is not returnable.
View return policy
Description
PDIA6 Polyclonal antibody specifically detects PDIA6 in Human,Mouse samples. It is validated for Immunohistochemistry,Western Blot
Specifications
| PDIA6 | |
| Polyclonal | |
| Tris-citrate/phosphate buffer, pH 7 to 8 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Immunohistochemistry, Western Blot | |
| Unconjugated | |
| EC 5.3.4.1, Endoplasmic reticulum protein 5, ERp5, ERP5ER protein 5, P5TXNDC7, protein disulfide isomerase family A, member 6, Protein disulfide isomerase P5, protein disulfide isomerase-associated 6, protein disulfide isomerase-related protein, protein disulfide-isomerase A6, thioredoxin domain containing 7 (protein disulfide isomerase), Thioredoxin domain-containing protein 7 | |
| Between 250 and 300 | |
| 100 μg | |
| Signal Transduction | |
| 10130 | |
| Store at 4°C. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction