missing translation for 'onlineSavingsMsg'
Learn More

PDIA6 Antibody (3B4), Novus Biologicals™

Product Code. 18332349 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18332349 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18332349 Supplier Novus Biologicals Supplier No. H00010130M04

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

PDIA6 Monoclonal antibody specifically detects PDIA6 in Human samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen PDIA6
Applications Western Blot, ELISA
Classification Monoclonal
Clone 3B4
Conjugate Unconjugated
Formulation 1x PBS, pH 7.4
Gene Accession No. AAH01312
Gene Alias EC 5.3.4.1, Endoplasmic reticulum protein 5, ERp5, ERP5ER protein 5, P5TXNDC7, protein disulfide isomerase family A, member 6, Protein disulfide isomerase P5, protein disulfide isomerase-associated 6, protein disulfide isomerase-related protein, protein disulfide-isomerase A6, thioredoxin domain containing 7 (protein disulfide isomerase), Thioredoxin domain-containing protein 7
Host Species Mouse
Immunogen PDIA6 (AAH01312, 1 a.a. ~ 440 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MALLVLGLVSCTFFLAVNGLYSSSDDVIELTPSNFNREVIQSDSLWLVEFYAPWCGHCQRLTPEWKKAATALKDVVKVGAVDADKHHSLGGQYGVQGFPTIKIFGSNKNRPEDYQGGRTGEAIVDAALSALRQLVKDRLGGRSGGYSSGKQGRSDSSSKKDVIELTDDSFDRNVLDSEDVWMVEFYAPWCGHCKNLEPEWAAAASEVKEQTKGKVKLAAVDATVNQVLASRYGIRGFPTIKIFQKGESPVDYDGGRTRSDIVSRALDLFSDNAPPPELLEIINEDIAKRTCEEHQLCVVAVLPHILDTGAAGRNSYLEVLLKLADKYKKKMWGWLWTEAGAQSELETALGIGGFGYPAMAAINARKMKFALLKGSFSEQGINEFLRELSFGRGSTAPVGGGAFPTIVEREPWDGRDGELPVEGDIDLSDVELDDLGKDEL
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 10130
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.