missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDIA2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 6 publications
£188.00 - £426.00
Specifications
| Antigen | PDIA2 |
|---|---|
| Dilution | Western Blot 1:10000-1:20000, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:5000-1:20000, Immunoprecipitation 1-3 ug, Immunohistochemistry-Paraffin 1:100-1:500, Immunohistochemistry-Frozen 1:100-1:1000 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18067073
|
Novus Biologicals
NB100-1921SS |
0.025 mL |
£188.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18709403
|
Novus Biologicals
NB100-1921 |
0.1 mL |
£426.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PDIA2 Polyclonal specifically detects PDIA2 in Human, Mouse, Rat, Bovine, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
| PDIA2 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cancer, Cellular Markers, ER Markers, Hypoxia | |
| Whole antisera with No Preservative | |
| 64714 | |
| Purified full length PDIA2 isolated from rat liver. | |
| Primary | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot 1:10000-1:20000, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:5000-1:20000, Immunoprecipitation 1-3 ug, Immunohistochemistry-Paraffin 1:100-1:500, Immunohistochemistry-Frozen 1:100-1:1000 | |
| Polyclonal | |
| Antisera | |
| RUO | |
| Human, Mouse, Rat, Bovine, Primate | |
| Pancreas-specific protein disulfide isomerase, pancreatic protein disulfide isomerase, PDA2, PDI, PDIp, PDIR, protein disulfide isomerase, pancreatic, protein disulfide isomerase-associated 2, protein disulfide-isomerase A2, Rho GDP dissociation inhibitor gamma | |
| PDIA2 | |
| IgG | |
| Unpurified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title