missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDHX Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-19749
This item is not returnable.
View return policy
Description
PDHX Polyclonal specifically detects PDHX in Human samples. It is validated for Western Blot.
Specifications
| PDHX | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.0) and 10% Glycerol with 0.01% Thimerosal | |
| Dihydrolipoamide dehydrogenase-binding protein of pyruvate dehydrogenasecomplex, DLDBP, E3-binding protein, E3BPpyruvate dehydrogenase complex, E3-binding protein subunit, OPDX, PDX1Lipoyl-containing pyruvate dehydrogenase complex component X, proXpyruvate dehydrogenase complex, lipoyl-containing component X, pyruvate dehydrogenase complex, component X, pyruvate dehydrogenase protein X component, mitochondrial | |
| Rabbit | |
| 54 kDa | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 8050 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.75mg/mL | |
| Western Blot 1:500-1:3000 | |
| O00330 | |
| PDHX | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human PDHX. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction