missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PDHB Polyclonal Antibody
Description
Recommended positive controls: A549, H1299, HCT116, mouse heart, rat heart. Predicted reactivity: Mouse (94%), Rat (94%), Zebrafish (85%), Xenopus laevis (84%), Pig (94%), Chicken (85%), Rhesus Monkey (100%), Chimpanzee (100%), Bovine (94%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
PDHB (Pyruvate dehydrogenase E1 component subunit beta, mitochondrial) is also named as PHE1B. The enzyme, which is found in mitochondria, is one of the component enzymes of the pyruvate dehydrogenase multienzyme complex (PDH). It catalyzes the first reaction of an oxidative decarboxylation sequence converting pyruvate to acetyl-CoA and CO(2). Defects in PDHB are the cause of pyruvate dehydrogenase E1-beta deficiency (PDHBD).
Specifications
Specifications
| Antigen | PDHB |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 |
| Gene | PDHB |
| Gene Accession No. | P11177, P49432, Q9D051 |
| Gene Alias | 2610103L06Rik; AL024199; C81408; PDHB; PDHBD; PDHE1-B; PHE1B; pyruvate dehydrogenase (lipoamide) beta; pyruvate dehydrogenase E1 component subunit beta, mitochondrial; pyruvate dehydrogenase, E1 beta polypeptide |
| Gene Symbols | PDHB |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?