missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PDHA1 Recombinant Rabbit Monoclonal Antibody (5N5W8)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA555220
This item is not returnable.
View return policy
Description
Immunogen Sequence: IPGLRVDGMD ILCVREATRF AAAYCRSGKG PILMELQTYR YHGHSMSDPG VSYRTREEIQ EVRSKSDPIM LLKDRMVNSN LASVEELKEI DVEVRKEIED.
The pyruvate dehydrogenase (PDH) complex is a nuclear-encoded mitochondrial multienzyme complex that catalyzes the overall conversion of pyruvate to acetyl-CoA and CO(2), and provides the primary link between glycolysis and the tricarboxylic acid (TCA) cycle. The PDH complex is composed of multiple copies of three enzymatic components: pyruvate dehydrogenase (E1), dihydrolipoamide acetyltransferase (E2) and lipoamide dehydrogenase (E3). The E1 enzyme is a heterotetramer of two alpha and two beta subunits. This gene encodes the E1 alpha 1 subunit containing the E1 active site, and plays a key role in the function of the PDH complex. Mutations in this gene are associated with pyruvate dehydrogenase E1-alpha deficiency and X-linked Leigh syndrome. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Specifications
| PDHA1 | |
| Recombinant Monoclonal | |
| 0.26 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| P08559, P35486 | |
| PDHA1 | |
| A synthetic peptide corresponding to a sequence within amino acids 248-348 of human PDHA1. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 5N5W8 | |
| Unconjugated | |
| PDHA1 | |
| PDHA; PDHA1; Pdha-1; PDHAD; PDHCE1A; PDHE1-A type I; PHE1A; pyruvate dehydrogenase; pyruvate dehydrogenase (lipoamide) alpha 1; pyruvate dehydrogenase alpha 1; pyruvate dehydrogenase complex, E1-alpha polypeptide 1; pyruvate dehydrogenase E1 alpha 1; pyruvate dehydrogenase E1 alpha 1 subunit; pyruvate dehydrogenase E1 component alpha subunit; pyruvate dehydrogenase E1 component alpha subunit precursor; pyruvate dehydrogenase E1 component subunit alpha, somatic form, mitochondrial; pyruvate dehydrogenase E1 subunit; pyruvate dehydrogenase E1 subunit alpha 1; pyruvate dehydrogenase E1alpha subunit | |
| Rabbit | |
| Protein A | |
| RUO | |
| 18597, 5160 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction