missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDH E1 alpha Mouse anti-Human, Clone: 2B3C10, Proteintech
Mouse Monoclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | PDH E1 alpha |
|---|---|
| Clone | 2B3C10 |
| Concentration | 1.87 mg/mL |
| Applications | Immunocytochemistry, Immunoprecipitation, Immunofluorescence, Flow Cytometry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16857705
|
Proteintech
66119-1-IG-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16847705
|
Proteintech
66119-1-IG-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Pyruvate dehydrogenase (PDH) is a mitochondrial multienzyme complex that catalyzes the oxidative decarboxylation of pyruvate and is one of the major enzymes responsible for the regulation of homeostasis of carbohydrate fuels in mammals. The enzymatic activity is regulated by a phosphorylation/dephosphorylation cycle. Phosphorylation of PDH by a specific pyruvate dehydrogenase kinase (PDK) results in inactivation.Specifications
| PDH E1 alpha | |
| 1.87 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| PDH E1alpha, PDHA1, PDHE1 A type I, pyruvate dehydrogenase E1 alpha subunit | |
| PDHA1 | |
| IgG2a | |
| Protein A | |
| Antibody |
| 2B3C10 | |
| Immunocytochemistry, Immunoprecipitation, Immunofluorescence, Flow Cytometry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Human | |
| P08559 | |
| 5160 | |
| PDH E1alpha Fusion Protein Ag12556 | |
| Primary | |
| -20°C | |
| PDHA1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title