missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PDGFRL Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA565741
This item is not returnable.
View return policy
Description
Immunogen sequence: RGFVYLQPHSE HQGVVYCRAE AGGRSQISVK YQLLYVAVPS GPPSTTILAS SNKVKSGDDI SVLCTVLGEP DVEVEFTWIF PGQKDERPVT IQDTWRLIHR GLGHTTRISQ SVITVEDFET IDAGYYICTA QNLQGQTTV Highest antigen sequence identity to the following orthologs - mouse 93%, rat 94%.
PDGFRL bears significant sequence similarity to the ligand binding domain of platelet-derived growth factor receptor beta. Mutations in the PDGFRL gene, or deletion of a chromosomal segment containing this gene, are associated with sporadic hepatocellular carcinomas, colorectal cancers, and non-small cell lung cancers. This suggests PDGFRL may function as a tumor suppressor.
Specifications
| PDGFRL | |
| Polyclonal | |
| Unconjugated | |
| PDGFRL | |
| 1110039P19Rik; AV013190; PDGF receptor beta-like tumor suppressor; PDGFRL; PDGFR-like protein; PDGRL; platelet derived growth factor receptor like; platelet-derived growth factor receptor-like; platelet-derived growth factor receptor-like protein; platelet-derived growth factor-beta-like tumor suppressor; PRLTS | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 5157 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q15198 | |
| PDGFRL | |
| Recombinant Human PDGFRL. Recombinant protein control fragment (Product #RP-106417). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction