missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDGFRL Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16217-1-AP-20UL
This item is not returnable.
View return policy
Description
PDGFRL bears significant sequence similarity to the ligand binding domain of platelet-derived growth factor receptor beta. Mutations in the PDGFRL gene, or deletion of a chromosomal segment containing this gene, are associated with sporadic hepatocellular carcinomas, colorectal cancers, and non-small cell lung cancers. This suggests PDGFRL may function as a tumor suppressor.Specifications
| PDGFRL | |
| Polyclonal | |
| Unconjugated | |
| PDGFRL | |
| PDGFR like protein, PDGFRL, PDGRL, PRLTS | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 290771, 5157, 68797 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Flow Cytometry, Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| 0.45 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q15198, Q5RJP7, Q6PE55 | |
| Pdgfrl | |
| PDGFRL Fusion Protein Ag9218 | |
| 20 μL | |
| Primary | |
| Mouse, Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction