missing translation for 'onlineSavingsMsg'
Learn More

PDGFRL Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Product Code. 18362007 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18362007 100 μL 100µL
18393097 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18362007 Supplier Novus Biologicals Supplier No. NBP304738100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PDGFRL Polyclonal specifically detects PDGFRL in Human, Mouse samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PDGFRL
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias PDGFR-like protein, PDGRL, platelet-derived growth factor receptor-like, platelet-derived growth factor receptor-like protein, platelet-derived growth factor-beta-like tumor suppressor, PRLTSPDGF receptor beta-like tumor suppressor
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 15-205 of human PDGFRL (NP_006198.1).
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5157
Target Species Human, Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.