missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PDGFRB Recombinant Rabbit Monoclonal Antibody (206), PE, Invitrogen™
Rabbit Recombinant Monoclonal Antibody
£93.50 - £283.00
Specifications
| Antigen | PDGFRB |
|---|---|
| Clone | 206 |
| Concentration | 100 μg/mL |
| Applications | Flow Cytometry |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30019602
|
Invitrogen™
MA552222 |
100 Tests |
£283.00
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30019265
|
Invitrogen™
MA552223 |
25 Tests |
£93.50
Pack of 25 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PDGFRB Recombinant Monoclonal antibody specifically detects PDGFRB in Human samples. It is validated for Flow CytometrySpecifications
| PDGFRB | |
| 100 μg/mL | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 0.5% BSA and 0.09% sodium azide | |
| AI528809; Beta platelet-derived growth factor receptor; beta-type platelet-derived growth factor receptor; CD140 antigen-like family member B; CD140B; CD140b antigen; IBGC4; IMF1; JTK12; KOGS; PDGF beta chain; PDGF Receptor beta; Pdgfr; Pdgfr1; PDGFR-1; PDGFRB; PDGF-R-beta; PDGFR-beta; PENTT; platelet derived growth factor receptor beta; platelet derived growth factor receptor, beta polypeptide; platelet-derived growth factor receptor 1; Platelet-derived growth factor receptor beta; platelet-derived growth factor receptor beta variant 1; Platelet-derived growth factor receptor, beta; platelet-derived growth factor receptor, beta polypeptide | |
| Recombinant Human PDGFRB/CD140b Protein (aa 1-531). | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Pdgfrb |
| 206 | |
| Flow Cytometry | |
| PE | |
| Rabbit | |
| Human | |
| P09619 | |
| 5159 | |
| IgG | |
| Protein A | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title