Learn More
Invitrogen™ PDGFRA [V561D] Recombinant Human Protein
PDGFRA is a cell surface receptor tyrosine kinase.
Brand: Invitrogen™ PV4681
Code : NEW
Additional Details : Weight : 0.01000kg
Descrizione
PDGFRA binds members of the platelet-derived growth factor family. The PDGFRA V561D mutation is observed in gastrointestinal stromal tumors (GIST).
Specifica
NP_006197, V561D | |
50mM Tris, pH 7.5, 150mM NaCl, 0.02% Triton X-100, 0.5mM EDTA, 2mM DTT, 50 % Glycerol | |
1mg | |
RUO - research use only | |
PDGFRA V561D, PDGFR alpha | |
Human | |
GST Tag | |
Frozen |
Nuclear Receptor Assay | |
89.8kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Cytoplasmic | |
PDGFR Kinase Family | |
Recombinant | |
Baculovirus |