missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PDGFRA [T674I] Recombinant Human Protein
PDGFRA T674I is a Gleevec resistant form of PDGFRA.
Brand: Invitrogen™ PV4118
Code : NEW
Additional Details : Weight : 0.00900kg
Description
PDGFRA is a cell surface receptor tyrosine kinase which binds members of the platelet-derived growth factor family.
Specifications
NP_006197, T674T | |
50mM Tris-HCl, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol | |
1mg | |
RUO - research use only | |
PDGFRA T674I, PDGFR alpha | |
Human | |
GST Tag | |
Frozen |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
89.8kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Cytoplasmic | |
PDGFR Kinase Family | |
Recombinant | |
Baculovirus |