missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDGFRA Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA516742
This item is not returnable.
View return policy
Description
PA5-16742 targets PDGFR alpha in IHC (P) applications and shows reactivity with Human and mouse samples. The PA5-16742 immunogen is a synthetic peptide derived from C-terminal of human PDGFR alpha.
PDGFR (Platelet Derived Growth Factor Receptor) exhibits two different isoforms, alpha; and beta; coded by two different genes. PDGF A receptors are expressed in oligodendrocyte progenitor cells and PDGF B receptors are expressed on neurons. This antibody is specific for PDGFR, alpha; and does not react with PDGFR, beta;.
Specifications
| PDGFRA | |
| Polyclonal | |
| PBS with 1% BSA and 0.1% sodium azide; pH 7.6 | |
| P16234, P26618 | |
| PDGFRA | |
| A synthetic peptide derived from C-terminal of human PDGFR alpha | |
| 500 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PDGFRA | |
| PDGF A-chain, PDGF subunit A, PDGF-1, platelet-derived growth factor A chain, platelet-derived growth factor alpha chain, platelet-derived growth factor alpha isoform 2 preproprotein, platelet-derived growth factor subunit A, pdgfr-a | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 18595, 5156 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction