missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDGFR alpha Antibody (OTI1C10), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73303H
This item is not returnable.
View return policy
Description
PDGFR alpha Monoclonal specifically detects PDGFR alpha in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| PDGFR alpha | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| PDGFRA | |
| Recombinant protein fragment corresponding to amino acids 789-1089 of human PDGF R alpha (NP_006197) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI1C10 | |
| Western Blot, Immunohistochemistry | |
| alpha-type platelet-derived growth factor receptor, CD140 antigen-like family member A, CD140a, CD140a antigen, EC 2.7.10, EC 2.7.10.1, MGC74795, PDGFR2, PDGFRA/BCR fusion, PDGF-R-alpha, platelet-derived growth factor receptor, alpha polypeptide, rearranged-in-hypereosinophilia-platelet derived growth factor receptor alphafusion protein, RHEPDGFRA | |
| Mouse | |
| Affinity Purified | |
| Phospho Specific, Protein Kinase | |
| 5156.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction