missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDGFR alpha Antibody (APA5), PE/Cy5.5, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP1-43350PECY55
This item is not returnable.
View return policy
Description
PDGFR alpha Monoclonal specifically detects PDGFR alpha in Human, Mouse samples. It is validated for Flow Cytometry.
Specifications
| PDGFR alpha | |
| Monoclonal | |
| PE-Cyanine5.5 | |
| PBS with 0.05% Sodium Azide | |
| PDGFRA | |
| The immunogen for this antibody was CD140a. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| IgG2a κ |
| Flow Cytometry | |
| APA5 | |
| Flow Cytometry | |
| alpha-type platelet-derived growth factor receptor, CD140 antigen-like family member A, CD140a, CD140a antigen, EC 2.7.10, EC 2.7.10.1, MGC74795, PDGFR2, PDGFRA/BCR fusion, PDGF-R-alpha, platelet-derived growth factor receptor, alpha polypeptide, rearranged-in-hypereosinophilia-platelet derived growth factor receptor alphafusion protein, RHEPDGFRA | |
| Rat | |
| Protein A or G purified | |
| Phospho Specific, Protein Kinase | |
| 5156.0 | |
| Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction