missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDGFR alpha Antibody (APA5) - BSA Free, Novus Biologicals™
Rat Monoclonal Antibody
£245.00 - £397.00
Specifications
| Antigen | PDGF R alpha |
|---|---|
| Clone | APA5 |
| Concentration | 0.5 mg/mL |
| Dilution | Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18262461
|
Novus Biologicals
NBP1-43350-0.05MG |
0.05 mg |
£245.00
0.05mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18016584
|
Novus Biologicals
NBP1-43350-0.1MG |
0.1 mg |
£397.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PDGFR alpha Monoclonal specifically detects PDGFR alpha in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.Specifications
| PDGF R alpha | |
| 0.5 mg/mL | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen) | |
| Unconjugated | |
| RUO | |
| Mouse | |
| P26618 | |
| 5156 | |
| The immunogen for this antibody was CD140a. | |
| Primary | |
| Store at 4C. Do not freeze. |
| APA5 | |
| Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
| Monoclonal | |
| Rat | |
| Protein Kinase | |
| Aqueous, may contain carrier protein/stabilizer. with 0.09% Sodium Azide | |
| alpha-type platelet-derived growth factor receptor, CD140 antigen-like family member A, CD140a, CD140a antigen, EC 2.7.10, EC 2.7.10.1, MGC74795, PDGFR2, PDGFRA/BCR fusion, PDGF-R-alpha, platelet-derived growth factor receptor, alpha polypeptide, rearranged-in-hypereosinophilia-platelet derived growth factor receptor alphafusion protein, RHEPDGFRA | |
| PDGFRA | |
| IgG2a κ | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title