missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
PDGFR alpha Monoclonal antibody specifically detects PDGFR alpha in Human samples. It is validated for Immunohistochemistry,Western Blot,Neutralization
Specifications
Specifications
| Antigen | PDGFR alpha |
| Applications | Immunohistochemistry, Western Blot, Neutralization |
| Classification | Monoclonal |
| Clone | 35248 |
| Conjugate | APC |
| Formulation | PBS |
| Gene Alias | alpha-type platelet-derived growth factor receptor, CD140 antigen-like family member A, CD140a, CD140a antigen, EC 2.7.10, EC 2.7.10.1, MGC74795, PDGFR2, PDGFRA/BCR fusion, PDGF-R-alpha, platelet-derived growth factor receptor, alpha polypeptide, rearranged-in-hypereosinophilia-platelet derived growth factor receptor alphafusion protein, RHEPDGFRA |
| Host Species | Mouse |
| Immunogen | S. frugiperda insect ovarian cell line Sf 21-derived recombinant human PDGF R alpha |
| Purification Method | Protein A or G purified from hybridoma culture supernatant |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?