missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDGFR alpha Antibody (16A1), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-44581AF594
This item is not returnable.
View return policy
Description
PDGFR alpha Monoclonal specifically detects PDGFR alpha in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Flow (Cell Surface).
Specifications
| PDGFR alpha | |
| Monoclonal | |
| Alexa Fluor 594 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| PDGFRA | |
| CD140a-transfected NIH 3T3 cells | |
| 0.1 mL | |
| Primary | |
| The mouse monoclonal antibody 16A1 recognizes CD140a/ PDGF-RA, the 170 kDa alpha chain of platelet-derived growth factor receptor, which is widely expressed on a variety of mesenchymal-derived cells and plays pro-proliferative or anti-proliferative roles in various tumours. HLDA VI.; WS Code E022 | |
| Store at 4°C in the dark. |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| 16A1 | |
| Flow Cytometry, Immunocytochemistry/Immunofluorescence, Flow (Cell Surface) | |
| alpha-type platelet-derived growth factor receptor, CD140 antigen-like family member A, CD140a, CD140a antigen, EC 2.7.10, EC 2.7.10.1, MGC74795, PDGFR2, PDGFRA/BCR fusion, PDGF-R-alpha, platelet-derived growth factor receptor, alpha polypeptide, rearranged-in-hypereosinophilia-platelet derived growth factor receptor alphafusion protein, RHEPDGFRA | |
| Mouse | |
| Protein A purified | |
| Phospho Specific, Protein Kinase | |
| 5156.0 | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction