Learn More
Eurofins DiscoverX PDGFR α (550-end, V561D) Protein, Active
Description
Receptors that are tyrosine protein kinases include those receptors for platelet-derived growth factor (PDGF), insulin, epidermal growth factor (EGF) and certain lymphokines. Receptor tyrosine kinases are cell surface transmembrane proteins responsible for intracellular signal transduction. They are expressed in several cell types and, after activation by growth factor binding, trigger a series of intracellular pathways leading to a wide variety of responses; e.g. differentiation, proliferation, migration and invasion, angiogenesis, survival. Inhibition of PDGFA, Alpha platelet-derived growth factor receptor, PDGF-R-alpha, CD140a antigen) receptor kinase may be useful in the treatment of several tumors.
GenBank M21574
Specifications
Specifications
| Formulation | mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 300 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution. |
| Molecular Weight (g/mol) | 63.5 kDa |
| Quantity | 10 μg |
| Species | Human |
| Recombinant | Recombinant |
| Protein Tag | N-terminal 6His |
| Expression System | Recombinant enzyme expressed in Sf21 insect cells |
| Protein Form | Truncated |
| Purity or Quality Grade | ≥62% |
| Protein | PDGFRα (hu,550-end,V561D) active |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.