missing translation for 'onlineSavingsMsg'
Learn More

PDGF R beta, p Tyr740 Antibody, Novus Biologicals™

Product Code. 18335044 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18335044 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18335044 Supplier Novus Biologicals Supplier No. NBP313269

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PDGF R beta Polyclonal antibody specifically detects PDGF R beta in Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen PDGF R beta
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS, 1% BSA, 20% Glycerol (pH7)
Gene Alias beta-type platelet-derived growth factor receptor, CD140 antigen-like family member B, CD140b, CD140b antigen, EC 2.7.10, EC 2.7.10.1, JTK12, PDGFR, PDGFR1, PDGF-R-beta, platelet-derived growth factor receptor, beta polypeptide
Host Species Rabbit
Immunogen Carrier-protein conjugated synthetic peptide corresponding to residues around human PDGF R beta (phospho Tyr740). The exact sequence is proprietary.
Purification Method Antigen Affinity-purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Oncogenes, Phospho Specific, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 5159
Target Species Mouse
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.