missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDGF R beta [p Tyr1021] Antibody (JE48-54), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-32786
This item is not returnable.
View return policy
Description
PDGF R beta Monoclonal antibody specifically detects PDGF R beta in Human,Mouse,Rat samples. It is validated for Western Blot
Specifications
| PDGF R beta | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000 | |
| beta-type platelet-derived growth factor receptor, CD140 antigen-like family member B, CD140b, CD140b antigen, EC 2.7.10, EC 2.7.10.1, JTK12, PDGFR, PDGFR1, PDGF-R-beta, platelet-derived growth factor receptor, beta polypeptide | |
| Synthetic phospho-peptide corresponding to residues surrounding Tyr 1021 of human PDGF R beta. (Uniprot: P05622) | |
| 100 μL | |
| Oncogenes, Phospho Specific, Protein Kinase | |
| 5159 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| JE48-54 | |
| Unconjugated | |
| 1*TBS (pH7.4), 0.05% BSA and 40% Glycerol | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction