missing translation for 'onlineSavingsMsg'
Learn More

PDGF R beta Antibody (APB5), mFluor Violet 610 SE, Novus Biologicals™

Product Code. 18426922 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18426922 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18426922 Supplier Novus Biologicals Supplier No. NBP143349MFV610

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

PDGF R beta Monoclonal antibody specifically detects PDGF R beta in Mouse samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen PDGF R beta
Applications Flow Cytometry
Classification Monoclonal
Clone APB5
Conjugate mFluor Violet 610 SE
Dilution Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias beta-type platelet-derived growth factor receptor, CD140 antigen-like family member B, CD140b, CD140b antigen, EC 2.7.10, EC 2.7.10.1, JTK12, PDGFR, PDGFR1, PDGF-R-beta, platelet-derived growth factor receptor, beta polypeptide
Host Species Rat
Immunogen The immunogen for this antibody was PDGF receptor beta.
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Oncogenes, Phospho Specific, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 5159
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.