missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDGF R beta Antibody (18A2), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-47232JF646
This item is not returnable.
View return policy
Description
PDGF R beta Monoclonal specifically detects PDGF R beta in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications
| PDGFRbeta | |
| Monoclonal | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| PDGFRB | |
| CD140b-transfected NIH 3T3 cells | |
| 0.1 mL | |
| Primary | |
| The monoclonal antibody 18A2 recognizes CD140b PDGF-RB, the 180-190 kDa beta chain of platelet-derived growth factor receptor, which is widely expressed on a variety of mesenchymal-derived cells and plays pro-proliferative roles. | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| 18A2 | |
| Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
| beta-type platelet-derived growth factor receptor, CD140 antigen-like family member B, CD140b, CD140b antigen, EC 2.7.10, EC 2.7.10.1, JTK12, PDGFR, PDGFR1, PDGF-R-beta, platelet-derived growth factor receptor, beta polypeptide | |
| Mouse | |
| Protein A purified | |
| Oncogenes, Protein Kinase | |
| 5159 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction