missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDGF R beta Antibody (18A2), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-47232AF647
This item is not returnable.
View return policy
Description
PDGF R beta Monoclonal specifically detects PDGF R beta in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications
| PDGF R beta | |
| Monoclonal | |
| Alexa Fluor 647 | |
| beta-type platelet-derived growth factor receptor, CD140 antigen-like family member B, CD140b, CD140b antigen, EC 2.7.10, EC 2.7.10.1, JTK12, PDGFR, PDGFR1, PDGF-R-beta, platelet-derived growth factor receptor, beta polypeptide | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| The monoclonal antibody 18A2 recognizes CD140b PDGF-RB, the 180-190 kDa beta chain of platelet-derived growth factor receptor, which is widely expressed on a variety of mesenchymal-derived cells and plays pro-proliferative roles. | |
| Store at 4C in the dark. | |
| IgG1 |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| 18A2 | |
| Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
| PDGFRB | |
| CD140b-transfected NIH 3T3 cells | |
| 0.1 mL | |
| Phospho Specific, Protein Kinase | |
| 5159 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction