missing translation for 'onlineSavingsMsg'
Learn More

PDGF-D/SCDGFB Antibody (680018) [Janelia Fluor™ 525], Novus Biologicals™

Product Code. 30734120 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30734120 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30734120 Supplier Novus Biologicals Supplier No. FAB1159JF525

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

PDGF-D/SCDGFB Monoclonal antibody specifically detects PDGF-D/SCDGFB in Human samples. It is validated for Neutralization
TRUSTED_SUSTAINABILITY

Specifications

Antigen PDGF-D/SCDGFB
Applications Neutralization
Classification Monoclonal
Clone 680018
Conjugate Janelia Fluor 525
Formulation 50mM Sodium Borate
Gene Alias IEGFMSTP036, Iris-expressed growth factor, PDGF-D, platelet derived growth factor D, SCDGF-BMGC26867, SCDGFBplatelet-derived growth factor D, spinal cord derived growth factor B, Spinal cord-derived growth factor B, spinal cord-derived growth factor-B
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human PDGF-DD, Ser250-Arg370, Accession # Q9GZP0.1
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Vision
Primary or Secondary Primary
Gene ID (Entrez) 80310
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.