missing translation for 'onlineSavingsMsg'
Learn More

PDGF-C Antibody (619346) [DyLight 550], Novus Biologicals™

Product Code. 30737321 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30737321 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30737321 Supplier Novus Biologicals Supplier No. FAB1560L

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

PDGF-C Monoclonal antibody specifically detects PDGF-C in Human samples. It is validated for Neutralization
TRUSTED_SUSTAINABILITY

Specifications

Antigen PDGF-C
Applications Neutralization
Classification Monoclonal
Clone 619346
Conjugate DyLight 550
Formulation 50mM Sodium Borate
Gene Alias FALLOTEIN, PDGF-C, platelet derived growth factor C, platelet-derived growth factor C, SCDGFSpinal cord-derived growth factor, secretory growth factor-like protein, VEGF-E
Host Species Mouse
Immunogen E.coli-derived recombinant human PDGF-C, Val235-Gly345, Accession # Q9NRA1
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 56034
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.