missing translation for 'onlineSavingsMsg'
Learn More

PDGF-B Mouse anti-Human, Clone: PDGFB/3071, Novus Biologicals™

Product Code. 18399991 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
20 μg
Unit Size:
20µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18399991 20 μg 20µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18399991 Supplier Novus Biologicals Supplier No. NBP30781020UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

PDGF-B Monoclonal specifically detects PDGF-B in Human samples. It is validated for Immunohistochemistry-Paraffin, Protein Array.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PDGF-B
Applications Immunohistochemistry (Paraffin), Peptide Array
Classification Monoclonal
Clone PDGFB/3071
Concentration 0.2 mg/ml
Conjugate Unconjugated
Dilution Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array
Formulation 10 mM PBS with 0.05% BSA
Gene Alias B chain, Becaplermin, FLJ12858, PDGF subunit B, PDGF2, PDGF-2, platelet-derived growth factor 2, Platelet-derived growth factor beta polypeptide, platelet-derived growth factor beta polypeptide (simian sarcoma viral (v-sis)oncogene homolog), platelet-derived growth factor subunit B, platelet-derived growth factor, B chain, Platelet-derived growth factor, beta polypeptide (oncogene SIS), Proto-oncogene c-Sis, SSV
Host Species Mouse
Immunogen Recombinant fragment of human PDGF-B protein (around aa 27-158) (exact sequence is proprietary)
Purification Method Protein A or G purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Cell Cycle and Replication, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 5155
Target Species Human
Content And Storage Store at 4°C.
Form Purified
Isotype IgG2c κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.