missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDGF-B Antibody (S06-7H6), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-19733-50ul
This item is not returnable.
View return policy
Description
PDGF-B Monoclonal antibody specifically detects PDGF-B in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| PDGF-B | |
| Monoclonal | |
| 0.3 mg/mL | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| S06-7H6 | |
| Unconjugated | |
| B chain, Becaplermin, FLJ12858, PDGF subunit B, PDGF2, PDGF-2, platelet-derived growth factor 2, Platelet-derived growth factor beta polypeptide, platelet-derived growth factor beta polypeptide (simian sarcoma viral (v-sis)oncogene homolog), platelet-derived growth factor subunit B, platelet-derived growth factor, B chain, Platelet-derived growth factor, beta polypeptide (oncogene SIS), Proto-oncogene c-Sis, SSV | |
| A synthetic peptide of human PDGF-B (Uniprot # P01127) | |
| 50 μg | |
| Biologically Active Proteins, Cell Cycle and Replication, Stem Cell Markers | |
| 5155 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction