missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDGF-B Antibody (PDGFB/3072), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-08823CL1
This item is not returnable.
View return policy
Description
PDGF-B Monoclonal antibody specifically detects PDGF-B in Human samples. It is validated for Immunohistochemistry (Paraffin),Protein Array
Specifications
| PDGF-B | |
| Monoclonal | |
| CoraFluor 1 | |
| B chain, Becaplermin, FLJ12858, PDGF subunit B, PDGF2, PDGF-2, platelet-derived growth factor 2, Platelet-derived growth factor beta polypeptide, platelet-derived growth factor beta polypeptide (simian sarcoma viral (v-sis)oncogene homolog), platelet-derived growth factor subunit B, platelet-derived growth factor, B chain, Platelet-derived growth factor, beta polypeptide (oncogene SIS), Proto-oncogene c-Sis, SSV | |
| Recombinant fragment of human PDGF-B protein (around aa 27-158) (exact sequence is proprietary) (Uniprot: P01127) | |
| 0.1 mL | |
| Biologically Active Proteins, Cell Cycle and Replication, Stem Cell Markers | |
| 5155 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2c κ |
| Immunohistochemistry (Paraffin), Protein Array | |
| PDGFB/3072 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction