missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDF Antibody (OTI3F3), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73301PE
This item is not returnable.
View return policy
Description
PDF Monoclonal specifically detects PDF in Human samples. It is validated for Flow Cytometry.
Specifications
| Monoclonal | |
| PE | |
| PBS with 0.05% Sodium Azide | |
| Full length recombinant protein of human PDF (NP_071736) produced in E.coli. | |
| 0.1 mL | |
| 64146.0 | |
| Store at 4°C in the dark. |
| Flow Cytometry | |
| OTI3F3 | |
| Flow Cytometry | |
| EC 3.5.1.88, GDF15, MIC1, PDF1A, peptide deformylase (mitochondrial), peptide deformylase, mitochondrial, peptide deformylase-like protein, PLAB, Polypeptide deformylase | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction